Key Documents
BR91212
BRAND® glass beaker with spout, low form
volume 10 mL, without graduation
Synonym(s):
Beakers, Glass
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
material
borosilicate glass 3.3
feature
without graduation
manufacturer/tradename
BRAND 91212
volume
10 mL
General description
The BRAND® Glass Beaker with Spout is expertly crafted from Boro 3.3 glass, making it a reliable choice for laboratory applications. Designed in accordance with DIN EN ISO 3819 standards. The integrated spout allows for easy pouring, minimizing spills and enhancing accuracy during liquid transfer. (no graduations)
Other Notes
Browse all glass beakers
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Customers Also Viewed
Slide 1 of 1