MilliporeSigma
BR782871
BRAND® insert strips for 24 x 6-well BRANDplates®
Inlet Opening System, PET membrane, pore size 8 μm
Synonym(s):
BRANDplates®, Cell culture, Insert Strips, Insert System, Inserts, Tissue culture
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.22
UNSPSC Code:
41122107
Material:
PET membrane
Pore size:
8 μm
Sterility:
sterile
Binding type:
non-treated surface
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
PET membrane
description
cellGrade plus
sterility
sterile
manufacturer/tradename
BRAND 782871
packaging
pack of 12 ea (strips of 4 inserts, individually wrapped), pkg of (individually wrapped)
size
6 well
pore size
8 μm
binding type
non-treated surface
General description
Innovative cell culture plates and inserts for manual and automatic cell and tissue culture. The BRANDplates® Insert System, developed in conjunction with the Fraunhofer company, offers an innovative expansion to BRAND′s existing cell culture product range. It consists of two different cell culture plates and two designs of cell culture inserts that can be combined in different configurations.Insert specification:4x insert strips, can be divided into up to 4 individual inserts - hanging position in wellHanging inserts – distance between the well base and insert membrane is 4 mmIndividually packaged strips inserted in thermoformed beakers with 6 strips each in the 6-well cell culture platecellGrade plus (hydrophilic cell culture-) surface.Smooth wall inserts: For standard applications like secretion studies, co-cultivations, chemotaxis tests, etc.PS cellGrade plus surface, sterile, strip of 4 inserts (divisible).BIO-CERT® CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, Pyrogens and non cytotoxic
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
cellGrade is a trademark of BRAND GMBH + CO KG
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)