Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BR781415

BRAND® 48-well PCR plates

no skirt, thin-walled, flexible, clear

Synonym(s):

PCR, PCR Plates, Plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NB.22
material:
clear
colorless wells
polypropylene
size:
48 well
volume:
0.2 mL
working volume:
200 μm
sterility:
non-sterile
binding type:
non-treated surface
feature:
skirt: no
no skirt
thin-walled, flexible

material

clear
colorless wells
polypropylene

description

D-Nase/R-Nase, DNA-free, suitable for qPCR

sterility

non-sterile

feature

skirt: no
no skirt
thin-walled, flexible

packaging

pack of 20 ea

manufacturer/tradename

BRAND 781415

technique(s)

PCR: suitable

size

48 well

volume

0.2 mL

working volume

200 μm

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

These plates reduce material costs and allow work on a compact PCR plate even with small sample throughputs. The ultra thin-walled PCR plate design facilitates constant, rapid and precise heat transfer. The smooth vessel interior minimizes the binding of enzymes and nucleic acid to the walls. The raised lips of the wells protect against cross-contamination, allowing a reliable seal with the sealing mat tailored to the plates.

Features and Benefits

• Fits all common thermal cycler• Raised well edges protect against cross-contamination• Rapid heat transfer through thin vessel walls

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service