Key Documents
BR759015
BRAND® semi-micro cuvette
polystyrene, minimum filling volume 1.5mL
Synonym(s):
Cuvettes
About This Item
UNSPSC Code:
41121813
NACRES:
SB.32
material
polystyrene
packaging
pack of 100 ea
manufacturer/tradename
BRAND 759015
General description
  • BRAND® semi-micro cuvette is manufactured under controlled room conditions.
  • Clear, clean optical path with an indication of optical path orientation.
  • Packed grouped by mold cavity number to ensure the lowest variation of extinction coefficient.
  • Standard 10mm light path.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)