Key Documents
BR719005
BRAND® counting chamber BLAUBRAND® Malassez pattern
without clips, double ruled
Synonym(s):
BLAUBRAND®, Cell culture, Counting Chambers, Malassez
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
material
colorless
glass
sterility
non-sterile
feature
without clips
manufacturer/tradename
BRAND 719005
wells
1
General description
Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.The ruling is rectangular, covering an area of 5 mm². The large rectangles measure 0.25 x 0.20 = 0.05 mm². They are each subdivided into 20 mini squares with an area of 0.0025 mm².This chamber is commonly used for counting cells in the cerebro-spinal fluid, or for counting nematodes.
Features and Benefits
  • BLAUBRAND® quality
  • Outstanding measurement precision
  • 100% certified quality
Packaging
1 counting chamber, complete with 2 haemacytometer cover glasses, in transparent plastic box.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)