Key Documents
BR708214
Filling tube for Dispensette® S Organic / S Organic
1, 2, 5 and 10 mL, FEP tube, L 195-350 mm, individual adjustment of length
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
FEP tube
manufacturer/tradename
BRAND 708214
packaging
1 ea of
L
195-350 mm
O.D.
6 mm
capacity
1, 2, 5 and 10 mL
General description
Adjusts to various bottle heights.
Legal Information
Dispensette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)