Key Documents
BR707533
BRAND® dispenser cylinder with valve block for Titrette®
for 10 mL
Synonym(s):
Accessories, Bottle-top Burette, Burettes, Titrette®
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
packaging
pack of 1 ea
manufacturer/tradename
BRAND 707533
General description
Structural change from serial-number 01K (January 2012).The assembly of the titrating tube to the valve block has been changed. When ordering spare parts, the serial number must be accounted for.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Titrette is a registered trademark of BRAND GMBH + CO KG