Skip to Content
MilliporeSigma
All Photos(1)

Documents

BR705870

BRAND® Transferpette® S Pipette

tip volume (0.5-10 μL), single-channel, adjustable volume, mechanical, DE-M

Synonym(s):

air displacement pipette, microliter pipette, microliter pipettor, micropipette, micropipettor, pipette, pipettor, transferpette s

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.01

feature

single-channel adjustable volume
mechanical

packaging

case of 1 ea

manufacturer/tradename

BRAND 705870

tip volume

(0.5-10 μL)

Looking for similar products? Visit Product Comparison Guide

General description

The Transferpette® S air displacement pipette allows you to work efficiently and ergonomically with both small and large volumes. Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series. Due to its perfect ergonomics, the Transferpette S provides a comfortable grip in any hand position, whether you are right- or left-handed, or have big or small hands. Changing your grip is not necessary, since the microliter pipette provides true one-hand operation: set the volume, pipette, eject the tip. The volume-change protection is also easy to operate with gloves on and protects your set volume against accidental adjustment. The Transferpette S also features Easy Calibration technology, which makes calibration a breeze. Calibration can be carried out without tools. A compatibility table shows which tips fit which device, so that you always order the correct pipette tips.

Application

Pipetting

Features and Benefits

  • Efficient: one-hand operation
  • Flexible: ergonomic design for all hand sizes
  • Reliable: volume-change protection prevents accidental adjustments


Limitations of use:
  • Operating temperature of instrument and reagent should be between +15 °C and +40 °C (59 °F to 104 °F)
  • Vapor pressure up to 500 mbar
  • Viscosity: 260 mPa s

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service