Skip to Content
BR704652
BRAND® filter for Transferpette® 5000 μL
Synonym(s):
Accessories, Air displacement pipettes, Air interface pipettes, Microliter pipettes, Multichannel pipettes, Pipettes, Piston-operated pipettes, Single channel pipettes
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.01
UNSPSC Code:
41121600
Packaging:
pack of 25 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pack of 25 ea
manufacturer/tradename
BRAND 704652
General description
Filter compatible with Transferpette, Transferpette Pro and Electronic 2ml, 2,5ml and 5mL.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)