Key Documents
BR704522
BRAND® discharge tube seripettor® /seripettor® pro
for 2 & 10 mL
Synonym(s):
Accessories, Bottle-top Dispenser, Dispenser, seripettor®
About This Item
UNSPSC Code:
41121500
NACRES:
NB.43
material
PTFE
packaging
pack of 1 ea
manufacturer/tradename
BRAND 704522
L
800 mm
General description
Tubing PTFE, coiled, length 800 mm, with safety handle. Not suited for peroxide.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG