Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BR474305

BRAND® transparent plastic staining tray

Holds 20 x slides, for use with transparent plastic staining trough

Synonym(s):

Microscopy, Staining, Staining Accessories

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
MA.01

material

polypropylene

feature

Holds 20 x slides
for use with transparent plastic staining trough

packaging

pack of 2 ea

manufacturer/tradename

BRAND 474305

Features and Benefits

  • BRAND® transparent plastic staining tray is ideal for various staining procedures including immunostaining.
  • Includes a lid to help maintain humidity and reduce evaporation.
  • The staining tray has a fold-down handle for easy and accurate staining.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service