Documents
BR36968
BRAND® volumetric flask BLAUBRAND® USP conformity certified
capacity 5 mL, neck joint: ST/NS 10/19, glass stopper
Synonym(s):
BLAUBRAND®, USP, Volumetric flasks
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
glass stopper
Joint
joint: ST/NS neck10/19
feature
accuracy class A
packaging
pack of 2
manufacturer/tradename
BRAND 36968
capacity
5 mL
General description
The BRAND® volumetric flask BLAUBRAND® USP conformity certified is made from high-quality Boro 3.3 glass and complies with DIN EN ISO 1042 standards. It is calibrated to contain (TC, In), ensuring accurate liquid measurement for a variety of laboratory applications. Each flask comes with a USP batch certificate, confirming its adherence to stringent quality and safety standards. This volumetric flask is an essential tool for precise analytical work, making it ideal for laboratories that require reliable and certified liquid handling solutions.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG