BR27005
BRAND® SILBERBRAND ETERNA Graduated pipette, Class B, Type 2
2 mL with 0.01 mL graduations, not suitable for cotton plugs
Synonym(s):
ETERNA, Graduated Pipettes, Pipettes, SILBERBRAND
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Recommended Products
material
AR-Glass
packaging
pack of 12 ea
manufacturer/tradename
BRAND 27005
measuring range
accuracy: 0.01 mL
capacity
2.0 mL
Related Categories
General description
The BRAND® SILBERBRAND ETERNA graduated pipette is a Class B, Type 2 instrument made from AR-Glas® and complies with DIN EN ISO 835 standards. Designed for precision, it is calibrated to deliver (TD, Ex), ensuring accurate liquid transfer for various laboratory applications. This pipette combines durability with functionality, making it a reliable choice for professionals requiring high-quality measurement tools. Its clear graduation markings facilitate easy reading, enhancing user efficiency.
Legal Information
AR-Glas is a registered trademark of Schott AG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)