Documents
BR144673
BRAND® Conical Joint Stopper
borosilicate glass 3.3, joint: 14/23, amber, honed
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
amber
borosilicate glass 3.3
Joint
joint: 14/23
feature
honed
manufacturer/tradename
BRAND 144673
Related Categories
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG