Key Documents
BR144535
BRAND® conical joint stopper, borosilicate glass
joint: ST/NS 29/32, semi-hollow
Synonym(s):
Conical joint stoppers, Stoppers
About This Item
UNSPSC Code:
41122500
NACRES:
NB.51
material
glass
octagonal borosilicate glass (grip)
Joint
joint: ST/NS29/32
description
octagonal grip
packaging
pack of 1 ea
manufacturer/tradename
BRAND 144535
General description
DIN 12252. Solid or semi-hollow, with octagonal grip.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)