Key Documents
BMSV1002
Sigma-Aldrich
Aspire Laboratory Aspirator Accessories
Handheld vacuum controller with single channel adapters
About This Item
UNSPSC Code:
41100000
NACRES:
NB.79
manufacturer/tradename
Benchmark Scientific V1002
Related Categories
General description
Accessories designed for use with Aspire laboratory aspirator.
Handheld vacuum controller with single channel adapters.
Features and Benefits
  • The handheld controller can hold 200-300 ul disposable pipette tips
  • The handheld controller and adapters are composed of polyoxymethylene (POM)
  • Autoclave-friendly ergonomic designed for prolonged handheld usage
  • The controller is compatible with serological pipettes and Pasteur pipettes
Legal Information
6538 is a trademark of American Type Culture Collection
Aspire is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1