Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BMSD2400REUK

Benchmark BeadBlaster 24 Refrigerated Microtube Homogenizer

AC/DC input 230 V, UK plug

Synonym(s):

bead homogenization, bead grinder,cell lysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41101504
NACRES:
NB.13

feature

refrigerated
to hold 24 x 2 mL

manufacturer/tradename

Benchmark Scientific D2400-R-E-UK

Looking for similar products? Visit Product Comparison Guide

General description

The BeadBlaster 24R Refrigerated Microtube Homogenizer completely lyses, grinds and homogenizes a broad variety of biological samples. A powerful, compressor based refrigeration system maintains chamber temperature and protects heat sensitive samples during homogenization. The stainless steel tube carrier subjects up to 24 samples to an optimized, high speed, 3D motion, producing high energy impacts between the samples and microbeads to release cellular contents. After processing samples can be centrifuged and the lysate removed with a pipette. Operating parameters are set and displayed digitally up to 50 homogenization profiles can be stored in memory. A mechanical lid lock prevents opening the lid while the BeadBlaster is in operation. A variety of bead materials and sizes are available for use with a wide range of sample types.

Application

Bead homogenization

Legal Information

BeadBlaster is a trademark of Benchmark Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service