Key Documents
BMSBSH50022B
Sigma-Aldrich
myBlock ll digital dry bath with 2 Quick-Flip blocks for tube
strips and plates, AC/DC input 115 V AC, US 3-pin plug
About This Item
UNSPSC Code:
41102404
NACRES:
NB.76
manufacturer/tradename
Benchmark Scientific BSH5002-2B
parameter
+5 °C temp. range (+105 °C)
5-105 °C temp. range
capacity
, holds 2 blocks
General description
myBlock series is a "next generation" product line that introduces several major advantages over traditional dry baths. They are the first digital dry baths to include advanced microprocessor controls, timed or continuous operation and a removable hinged lid. In addition, an optional external temperature feedback probe can be positioned directly inside a sample tube, providing real-time control and monitoring of actual sample temperature.
MyBlock′s transparent lid helps to promote better uniformity and minimize temperature stratification in the block by maintaining a warm air jacket layer. The hinged lid can be flipped open (to the rear), or easily removed (lifted off) to accommodate taller tubes.
  • Digital temperature control (set and walk away)
  • Hinged lid (improves uniformity & conserves energy)
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)