Key Documents
BAF199170150
Scienceware® vacuum aspirator collection system
capacity 3200 mL, AC/DC input 115 V AC
Synonym(s):
bottle aspirator system
About This Item
UNSPSC Code:
41100000
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F19917-0150
capacity
3200 mL
Related Categories
General description
Perform vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes. To use, simply insert a pipette tip, Pasteur pipette, or serological pipette into the collection tube and plug in to a 110/120V outlet. Self-contained system with bottle, tubing and vacuum pump is lightweight, compact and easy to use.
Features and Benefits
  • Built-in pump quickly aspirates fluids while maintaining a low noise level (76db at 1 meter) and minimal vibration
  • Ideal for fluid collection from tissue and/or cell culture, Petri dishes, plates, flasks and tubes
  • Chemical and corrosion resistant polypropylene collection bottle holds 3800 mL (1 gallon) fluid wastes and has a safety release vent to prevent vacuum implosion
  • A replaceable hydrophobic filter seals itself if it becomes wet protecting the pump motor from fluid contamination
  • Compact design will not crowd workspace whether working in a hood or at the bench
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)