Skip to Content
MilliporeSigma
All Photos(2)

Documents

BAF187940028

SP Bel-Art® Poxygrid® Slanted Tube Rack

to hold, 8 x 50 mL tubes

Synonym(s):

Angled rack, slanting tube holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.43

feature

to hold 8 x 50 mL tubes

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F18794-0028

H × L × width

4.75 in. (121 mm) × 6 in. (152 mm) × 4.75 in. (121 mm)

General description

Angled rack is useful for keeping tube caps and marked contents easily readable, frequently used reagents at an ergonomic angle or draining excess supernatant fluid from pellets.

Features and Benefits

Angled rack is useful for keeping tube caps and marked contents easily readable, frequently used reagents at an ergonomic angle or draining excess supernatant fluid from pellets.
  • Holds eight 50ml conical tubes or other vessels up to 30mm in diameter
  • Can be attached to the underside of a shelf using built in mounting points (hardware not included); keeps samples off the bench but immediately available
  • Steel wire is epoxy coated for long life and easy cleaning; unaffected by organic solvents, salts, weak acids and weak alkalis
  • Autoclavable at 121ºC (250ºF)

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP
Poxygrid is a registered trademark of SP Scienceware, a division of SP

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service