B81204
Bromosuccinic acid
98%
Synonym(s):
2-Bromobutanedioic acid, 2-Bromosuccinic acid, DL-Bromosuccinic acid, Monobromosuccinic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HOOCCH2CHBrCOOH
CAS Number:
Molecular Weight:
196.98
Beilstein:
1723556
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
powder
mp
161-163 °C (lit.)
SMILES string
OC(=O)CC(Br)C(O)=O
InChI
1S/C4H5BrO4/c5-2(4(8)9)1-3(6)7/h2H,1H2,(H,6,7)(H,8,9)
InChI key
QQWGVQWAEANRTK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
R H Yamada et al.
Biochimica et biophysica acta, 801(1), 151-154 (1984-09-07)
DL-Hydrazinosuccinic acid was synthesized by the reaction of DL-bromosuccinic acid with hydrazine. The compound strongly inhibited aspartate aminotransferase and gave 50% inhibition at 1.3 microM when added simultaneously with L-aspartate to an assay mixture containing enzyme. Incubation of the enzyme
M Blumrich et al.
Biochimica et biophysica acta, 1029(1), 1-12 (1990-11-02)
The present study indicates that SH-groups are essential for the uptake of [3H]taurocholate and [14C]cholate into isolated rat hepatocytes. Several sulfhydryl-modifying reagents viz. p-chloromercuribenzenesulfonate (PCMBS), N-ethylmaleimide (NEM), dithio-bis(5-nitropyridine) (DTNP), bromosuccinimide and HgCl2 inhibited uptake of bile acids in a concentration-dependent
Modification of three active site lysine residues in the catalytic subunit of aspartate transcarbamylase by D- and L-bromosuccinate.
A M Lauritzen et al.
The Journal of biological chemistry, 257(3), 1312-1319 (1982-02-10)
Contact dermatitis from alpha-bromomethylparotolylsulfone.
S Kerre et al.
Contact dermatitis, 33(2), 130-130 (1995-08-01)
Vincent Perkins et al.
Frontiers in microbiology, 11, 737-737 (2020-05-28)
The yeast Geotrichum candidum (teleomorph Galactomyces candidus) is inoculated onto mold- and smear-ripened cheeses and plays several roles during cheese ripening. Its ability to metabolize proteins, lipids, and organic acids enables its growth on the cheese surface and promotes the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service