Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00633

Sigma-Aldrich

Methyl (+)-PI Reagent

≥95%

Synonym(s):

1,3,2-Benzoxathiaphosphole, hexahydro-2,7a-dimethyl-5-(1-methylethenyl)-, 2-sulfide, (2R,3aR,5S,7aR)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19OPS2
CAS Number:
Molecular Weight:
262.37
UNSPSC Code:
12352005
NACRES:
NA.06

Quality Level

Assay

≥95%

form

powder

storage temp.

2-8°C

SMILES string

C[C@]1(O[P@](S2)(C)=S)[C@@]2([H])C[C@@H](C(C)=C)CC1

Application

Methyl (+)-PI Reagent is a methyl substituted analog of the air and moisture-tolerant (+-)-PI reagent developed in the Baran lab for stereospecific synthesis of chiral phosphines and methylphosphonate nucleotides.

Learn more about PI and PSI reagents

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongmin Xu et al.
Journal of the American Chemical Society, 142(12), 5785-5792 (2020-02-29)
Phosphorus Incorporation (PI, abbreviated Π) reagents for the modular, scalable, and stereospecific synthesis of chiral phosphines and methylphosphonate nucleotides are reported. Synthesized from trans-limonene oxide, this reagent class displays an unexpected reactivity profile and enables access to chemical space distinct

Articles

PI and PSI reagents are a platform of phosphate-based reagents for phosphorylation and phosphothiolation of nucleotides and peptides developed by the Baran lab.

PI and PSI reagents are a platform of phosphate-based reagents for phosphorylation and phosphothiolation of nucleotides and peptides developed by the Baran lab.

PI and PSI reagents are a platform of phosphate-based reagents for phosphorylation and phosphothiolation of nucleotides and peptides developed by the Baran lab.

PI and PSI reagents are a platform of phosphate-based reagents for phosphorylation and phosphothiolation of nucleotides and peptides developed by the Baran lab.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service