Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00422

Sigma-Aldrich

2-Formylphenyl sulfofluoridate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5FO4S
Molecular Weight:
204.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

description

Fp: >230°F

Quality Level

form

liquid

refractive index

n20/D 1.5086

density

1.4545 g/mL at 25 °C

SMILES string

O=S(OC1=C(C=O)C=CC=C1)(F)=O

InChI

1S/C7H5FO4S/c8-13(10,11)12-7-4-2-1-3-6(7)5-9/h1-5H

InChI key

SUOCJDDQQKVIKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Formylphenyl sulfofluoridate is an aryl fluorosulfate. It can be synthesized from the reaction between phenol and sulfuryl fluoride in the presence of triethylamine. It undergoes Sonogashira coupling reaction with phenyl acetylene.

Application

The following aryl fluorosulfate can be utilized as a cross-coupling partner in Suzuki-Miyaura reactions. The standard reaction conditions for these palladium-catalyzed C-C bond formations are ligand-free and can be performed in water, at room temperature and are not air sensitive.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiaobin Liang et al.
Organic letters, 17(8), 1942-1945 (2015-04-10)
Aryl fluorosulfates were prepared by a simple method and employed as coupling partners in the Suzuki-Miyaura reaction. The cross-coupling reactions were performed in water under air at room temperature without ligands or additives such as surfactants or phase-transfer reagents and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service