Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00374

Sigma-Aldrich

L-Fmoc-3-fluorophenylalanine

Synonym(s):

Fmoc-Phe(3-F)-OH, Fmoc-3-fluoro-L-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H20FNO4
CAS Number:
Molecular Weight:
405.42
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

color

beige

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

FC1=CC=CC(C[C@H](NC(OCC2C3=C(C4=C2C=CC=C4)C=CC=C3)=O)C(O)=O)=C1

InChI

1S/C24H20FNO4/c25-16-7-5-6-15(12-16)13-22(23(27)28)26-24(29)30-14-21-19-10-3-1-8-17(19)18-9-2-4-11-20(18)21/h1-12,21-22H,13-14H2,(H,26,29)(H,27,28)/t22-/m0/s1

InChI key

DWSDVARCJDOADL-QFIPXVFZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phenylalanine derivative was introduced in collaboration with Yu and coworkers in reflection to a methodology reported in C-H Activation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Yu program centers around the discovery of catalytic carbon–carbon and carbon–heteroatom bond forming reactions based on C–H activation. Target transformations are selected to enable 1) the use of simple and abundant starting materials such as aliphatic acids, amines and alcohols, and 2) disconnections that drastically shorten the synthesis of a drug molecule or a major class of biologically active compounds.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service