Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ALD00024

Sigma-Aldrich

2-(2-Propyn-1-yloxy)ethanesulfonyl fluoride

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7FO3S
Molecular Weight:
166.17
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

reaction suitability

reaction type: click chemistry

refractive index

n20/D 1.435

density

1.290 g/mL at 25 °C

SMILES string

O=S(CCOCC#C)(F)=O

InChI

1S/C5H7FO3S/c1-2-3-9-4-5-10(6,7)8/h1H,3-5H2

InChI key

HVQHLNVMEKMUEA-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service