Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00004

Sigma-Aldrich

Li-Yu t-Butyl Quinoline

95%

Synonym(s):

3,4-Dihydro-7-tert-butyl-2,5-dimethyl-2H-pyrano[2,3-b]quinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H23NO
Molecular Weight:
269.38
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

reaction suitability

reaction type: C-C Bond Formation
reagent type: catalyst
reagent type: ligand
reaction type: C-H Activation

mp

202-207 °C

SMILES string

CC1=C2C(OC(C)CC2)=NC3=C1C=C(C(C)(C)C)C=C3

InChI

1S/C18H23NO/c1-11-6-8-14-12(2)15-10-13(18(3,4)5)7-9-16(15)19-17(14)20-11/h7,9-11H,6,8H2,1-5H3

InChI key

HGKGTNXPFXRVQB-UHFFFAOYSA-N

Application

Ligand is optimal for gamma C-H functionalization of aliphatic acids through Pd-catalyzed C-H activation. The Yu group has demonstrated a variety of transformations, such as olefination, carbonylation, and arylation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Yu program centers around the discovery of catalytic carbon–carbon and carbon–heteroatom bond forming reactions based on C–H activation. Target transformations are selected to enable 1) the use of simple and abundant starting materials such as aliphatic acids, amines and alcohols, and 2) disconnections that drastically shorten the synthesis of a drug molecule or a major class of biologically active compounds.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service