Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

A68807

Sigma-Aldrich

4-Nitro-3-(trifluoromethyl)aniline

98%

Synonym(s):

4-Nitro-α,α,α-trifluoro-m-toluidine, 5-Amino-2-nitrobenzotrifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3(CF3)NH2
CAS Number:
Molecular Weight:
206.12
Beilstein:
2650702
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

125-129 °C (lit.)

SMILES string

Nc1ccc(c(c1)C(F)(F)F)[N+]([O-])=O

InChI

1S/C7H5F3N2O2/c8-7(9,10)5-3-4(11)1-2-6(5)12(13)14/h1-3H,11H2

InChI key

UTKUVRNVYFTEHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

M Schulz et al.
European journal of clinical pharmacology, 34(6), 633-636 (1988-01-01)
Flutamide is a nonsteroidal antiandrogen used in the treatment of prostatic carcinoma. We have investigated the disposition of flutamide and its two major metabolites in ten urological in-patients without significant liver or renal disease. After oral administration flutamide is absorbed
Gábor Benkovics et al.
International journal of pharmaceutics, 531(2), 614-620 (2017-05-16)
This contribution reports the design, synthesis and photochemical properties of a novel cationic, water soluble, β-cyclodextrin (βCD) conjugate integrating an anthracene moiety and a nitroaniline derivative within the primary side of the βCD scaffold. Photoinduced energy transfer between the anthracene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service