Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

940674

Sigma-Aldrich

DBCO-NHCO-PEG4-amine TFA

new

≥95%

Synonym(s):

15-Amino-N-[3-(11,12-didehydrodibenz[b,f]azocin-5(6H)-yl)-3-oxopropyl]-4,7,10,13-tetraoxapentadecanamide, 4,7,10,13-Tetraoxapentadecanamide, 15-amino-N-[3-(11,12-didehydrodibenz[b,f]azocin-5(6H)-yl)-3-oxopropyl], ADIBO-PEG4-amine TFA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H37N3O6 · xC2HF3O2
CAS Number:
Molecular Weight:
523.62 (free base basis)
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95%

form

solid

color

colorless to yellow-brown

storage temp.

−20°C

SMILES string

O=C(NCCC(=O)N1C=2C=CC=CC2C#CC=3C=CC=CC3C1)CCOCCOCCOCCOCCN

InChI

1S/C29H37N3O6.C2HF3O/c30-13-16-36-18-20-38-22-21-37-19-17-35-15-12-28(33)31-14-11-29(34)32-23-26-7-2-1-5-24(26)9-10-25-6-3-4-8-27(25)32;3-2(4,5)1-6/h1-8H,11-23,30H2,(H,31,33);1H

InChI key

MRRSYYMIRSTYMO-UHFFFAOYSA-N

Application

DBCO-PEG4-Amine TFA is a heterobifunctional building block with two reactive terminals. The Dibenzocyclooctyne (DBCO) terminus is useful for copper-free click chemistry. DBCO compounds react with azides without need of Cu(I) catalyst due to the strained nature of alkyne resulting in formation of fast and stable triazole linkage. The amine terminus is useful for reaction with carboxylic acid containing linkers in presence in actiavtors such as EDC or HATU.

Features and Benefits

The PEG4 linker is suited for improving solubility of resulting conjugates and provides sufficient length to minimize interactions between macromolecules of interest.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Click chemistry for the preparation of advanced macromolecular architectures
Akeroyd N, et al.
European Polymer Journal, 47, 1207-1231 (2010)
A stepwise huisgen cycloaddition process: copper(I)-catalyzed regioselective "ligation" of azides and terminal alkynes.
Vsevolod V Rostovtsev et al.
Angewandte Chemie (International ed. in English), 41(14), 2596-2599 (2002-08-31)
Pamela V Chang et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(5), 1821-1826 (2010-01-19)
Chemical reactions that enable selective biomolecule labeling in living organisms offer a means to probe biological processes in vivo. Very few reactions possess the requisite bioorthogonality, and, among these, only the Staudinger ligation between azides and triarylphosphines has been employed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service