Skip to Content
MilliporeSigma
All Photos(1)

Documents

939404

Sigma-Aldrich

tert-Butyllithium solution

new

1.7 M (in PAO/hexanes mixture)

Synonym(s):

t-BuLi, Lithium-2-methyl-2-propanide, t-BuLi

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CLi
CAS Number:
Molecular Weight:
64.06
Beilstein:
3587204
MDL number:
UNSPSC Code:
12352142

form

liquid

Quality Level

concentration

1.7 M (in PAO/hexanes mixture)

color

colorless to yellow

storage temp.

2-8°C

SMILES string

[Li]C(C)(C)C

InChI

1S/C4H9.Li/c1-4(2)3;/h1-3H3;

InChI key

BKDLGMUIXWPYGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butyllithium solution (tert-BuLi) is an organolithium compound, used as a strong base in organic chemistry. It facilitates lithium-halogen exchange reaction and also can be employed in the deprotonation of C-H compounds and amines.

Application

tert-Butyllithium solution can be used as a strong base in the synthesis of: Alkyllithium derivatives form alkyl iodides and diiodoalkanes by halogen-lithium exchange reaction. Inorganic polymer [(LiMo3Se3)n] from [(InMo3Se3)n] by reductive intercalation method. Adducts of cinnamic acid (1,3- and 1,4-adducts) by reacting with C-C double of cinnamic acid.

Features and Benefits

This product is formulated in a solution of polyalphaolefin (PAO) and hexanes to prevent pyrophoric reactions when exposed to air, affording improved safety and usability.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 1 Inhalation - Water-react 1

Target Organs

Nervous system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

88.7 °F

Flash Point(C)

31.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service