Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

936197

Sigma-Aldrich

AdQPhos Pd G3

Synonym(s):

AdQPhos G3 Palladacycle, AdQPhos Palladacycle

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C73H72FeNO3PPdS
Molecular Weight:
1236.66
UNSPSC Code:
12352142
NACRES:
NA.22

Assay

≥90%

Quality Level

form

powder

reaction suitability

reaction type: Cross Couplings

functional group

(palladium)
phosphine

SMILES string

CS(O[Pd]1([P](C23C[C@H]4C[C@H](C[C@H](C4)C3)C2)(C56C[C@H]7C[C@H](C[C@H](C7)C6)C5)c8cccc8)[NH2]C(C=CC=C9)=C9C%10=C1C=CC=C%10)(=O)=O.c%11(C%12=CC=CC=C%12)c(C%13=CC=CC=C%13)c(C%14=CC=CC=C%14)c(C%15=CC=CC=C%15)c%11C%16=CC=CC=C%16.[Fe]

General description

The precatalysts derived from AdQPhos exhibit excellent activity towards palladium-catalyzed α-arylation reactions under milder conditions. The strength of these catalytic systems in enabling such reactions were demonstrated with the broad scope of nucleophiles, including nitroalkanes, nitriles, amides, esters, and ketones. Compared to other state-of-the-art catalysts, this system was far superior and served as the first versatile catalytic system to accommodate all these nucleophiles in this context.

Further, by merging with micellar catalysis, this catalytic system demonstrated the feasibility of enabling α-arylation reactions under mild aqueous conditions. This approach provides an environmentally benign alternative to toxic/hazardous solvents like 1,4-dioxane or NMP, bringing modern organic synthesis closer to being ideally sustainable.

Application

In addition to facilitating α-arylations, these catalysts demonstrate compatibility with various challenging cross-coupling reactions, such as Suzuki-Miyaura, Buchwald-Hartwig aminations, and C-O and C-S cross-couplings.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service