Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

935921

Sigma-Aldrich

Poly(glycerol sebacate)

Synonym(s):

PGS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(OH)CH2OOC(CH2)8COO]n
UNSPSC Code:
12352201
NACRES:
NA.21

form

solid

Quality Level

color

colorless to yellow

General description

Poly(glycerol sebacate) (PGS) is a biodegradable polymer with tunable mechanical properties that can be used in soft tissue engineering and regenerative medicine applications. It is non-immunogenic, non-cytotoxic and has minimal inflammatory response. The polymer backbone and crosslinks can undergo hydrolytic degradation due to the presence of ester linkages. The ester bonds in the polymer backbone covalently crosslink at high temperature (>130 °C) under vacuum without using any toxic crosslinkers to form a three-dimensional (3D) network of random coils.

Application

  • Scaffold material synthesis like blood vessels, cornea and nerve tissue
  • In orthopedic implants
  • In the drug delivery system and bioelectronic applications

Features and Benefits

  • Flexible elastomer with tunable mechanical properties
  • Higher reversible deformation properties
  • Shape memory
  • Bioresorbable polymer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, Characterization and 3D Micro-Structuring via 2-Photon Polymerization of Poly(glycerol sebacate)-Methacrylate?An Elastomeric Degradable Polymer
Tala, et al.
Frontiers in physics, 6, 41-41 (2018)
Poly(Glycerol Sebacate) in Biomedical Applications-A Review of the Recent Literature
Vogt L, et al.
Advanced Healthcare Materials, 10, e2002026-e2002026 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service