Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

934674

Sigma-Aldrich

Activated carbon

greener alternative

Electrode sheet, copper substrate, size 5 in. × 10 in.

Synonym(s):

Activated carbon, Activated carbon anode

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NA.21

material

copper substrate

Quality Level

grade

battery grade

description

Description/Nominal Voltage: 0.3 V vs Li/Li+

Assay

≥98%

composition

Active material loading 5.7 mg/cm2 ± 5%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

size

5 in. × 10 in.

thickness

10 μm , copper current collector
90 μm , (excluding current collector)

particle size

5 -10 μm (active material characteristic)

capacity

220 mAh/g±5 % (Charge Capacity)

application(s)

battery manufacturing

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Activated carbon is used as anode in rechargeable batteries due to its many advantageous properties such as a very low insertion/deinsertion potential, large surface area, high pore density, good conductivity, low cost, and good mechanical and chemical stability compared to other alternatives. Experimentally it has been observed that by using activated carbon electrode, the diffusion co-efficient of potassium ion in potassium ion batteries improved by 7 times and could deliver a a high reversible capacity of 100 mAh/g after 100 cycles (at a high current density of 0.2 A/g), and exhibits increased rate performance. In addition, owing to the typical characteristic of porous structure, activated carbon (AC) is widely used for supercapacitor electrode and also possesses the potential as anode material in lithium ions batteries (LIBs).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coal-Derived Activated Carbon for Electrochemical Energy Storage: Status on Supercapacitor, Li-Ion Battery, and Li?S Battery Applications
Mousumi Bora
Energy & Fuels, 35, 18285?18307-18285?18307 (2021)
Activated carbon from the graphite with increased rate capability for the potassium ion battery.
Tai, Zhixin & Zhang
Carbon, 123, 54-61 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service