Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

933783

Sigma-Aldrich

Cobalt(II) oxide

greener alternative

nanopowder, 50 nm avg. part. size, ≥99.5% trace metals basis

Synonym(s):

Cobalt Monoxide, Oxocobalt, Cobaltous oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoO
CAS Number:
Molecular Weight:
74.93
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.21

Quality Level

Assay

≥99.5% trace metals basis

form

nanopowder

composition

CoO

storage condition

(Containers should be closed tightly and stored in a dry, cool and well-ventilated place.)

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

avg. part. size

50 nm

bulk density

0.966 g/cm3

application(s)

battery manufacturing

greener alternative category

SMILES string

O=[Co]

InChI

1S/Co.O

InChI key

IVMYJDGYRUAWML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our cobalt (II) oxide nanopowder is a high-quality, fine powder with an particle size of 50 nm, a high surface area, and a purity greater than 99.5% (<5000 ppm trace metals impurities). It is a dark gray powder that is stable at room temperature and has excellent chemical and thermal stability.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product has been enhanced for energy efficency. Click here for more information.

Application

Cobalt (II) oxide nanopowder is widely used in many industrial applications due to its unique physical and chemical properties. It is commonly used as a catalyst in chemical reactions such as the oxidation of hydrocarbons and the reduction of nitrogen oxides. In the field of energy storage, cobalt (II) oxide nanopowder is used in the fabrication of high-performance supercapacitors due to its high capacitance, fast charge-discharge characteristics, and excellent cycling stability. In addition, it is also used in the development of magnetic materials, including magnetic nanoparticles, and as a precursor for the synthesis of cobalt-based cathode materials for lithium-ion batteries.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Repr. 1B - Resp. Sens. 1B - Skin Sens. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiayu Chu et al.
Nanoscale, 11(33), 15633-15640 (2019-08-14)
In order to further enhance the performance of photocatalysts, cocatalysts are used to accelerate the photocatalytic reactions. Herein, ultrafine cobalt oxide (CoO) nanoparticles are synthesized through a novel bottom-up strategy and explored as an efficient non-noble cocatalyst to dramatically promote
Renata V Santos et al.
Scientific reports, 11(1), 21028-21028 (2021-10-27)
We herein report a comprehensive investigation on the magnetic, structural, and electric properties of CoO nanoparticles with different sizes by local inspection through hyperfine interactions measured in a wide range of temperatures (10-670 K) by using radioactive [Formula: see text]In([Formula:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service