Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

932884

Sigma-Aldrich

Tweezer

EMS 7 SA/Dr

Synonym(s):

Gripper

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

General description

EMS techware

Application

Safe ergonomic cushion grips (resistivity 10E9 Ohm) for handling ESD sensitive components or small static items. Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed.

OAL: 115 mm. Tips: Fine and curve.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service