Skip to Content
MilliporeSigma

930652

Pomalidomide-C2-NH2 hydrochloride

≥95%

Synonym(s):

4-((2-aminoethyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride, 4-[(2-Aminoethyl)amino]-2-(2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione, E3 Ligase Ligand 17

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H16N4O4 · xHCl
CAS Number:
Molecular Weight:
316.31 (free base basis)
UNSPSC Code:
12352101
NACRES:
NA.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


ligand

pomalidomide

Quality Segment

assay

≥95%

form

powder

functional group

amine

storage temp.

2-8°C

SMILES string

O=C1NC(C(CC1)N2C(C3=C(C2=O)C(NCCN)=CC=C3)=O)=O.Cl

InChI

1S/C15H16N4O4.ClH/c16-6-7-17-9-3-1-2-8-12(9)15(23)19(14(8)22)10-4-5-11(20)18-13(10)21;/h1-3,10,17H,4-7,16H2,(H,18,20,21);1H

InChI key

DSJCOXQYDMWPEY-UHFFFAOYSA-N

Application

Pomalidomide-C2-NH2 Hydrochloride is a functionalized cereblon ligand for development of pomalidomide-based PROTACs. Allows rapid conjugation with carboxyl linkers due to the presence of an amine group via peptide coupling reactions. It is also active for linker attachment via reductive amination, and a basic building block for development of a protein degrader library.


Technology Spotlight: Degrader Building Blocks for Targeted Protein Degradation


Protein Degrader Building Blocks

Legal Information

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library