Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

929948

Sigma-Aldrich

Ursodeoxycholic acid-22,23,24-13C3 solution

≥98 atom % 13C, ≥-98% (CP), 100 μg/mL in methanol

Synonym(s):

(R)-4-((3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[i]phenanthren-17-yl-2,3,4-13C3)pentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C21H40O4
Molecular Weight:
395.55
UNSPSC Code:
12142200
NACRES:
NA.13

isotopic purity

≥98 atom % 13C

Quality Level

Assay

≥-98% (CP)

concentration

100 μg/mL in methanol

mass shift

M+3

SMILES string

O[13C@@H]1[13CH2]C[C@@]2(C)[C@@](C[C@H](O)[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)CCC(O)=O)([H])[13CH2]1

InChI

1S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16-,17-,18+,19+,20+,22+,23+,24-/m1/s1/i8+1,12+1,16+1

InChI key

RUDATBOHQWOJDD-AQETUQINSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service