928771
Microdot tips
Stainless steel, lavender, 30G, 0.25"
Synonym(s):
3D bioprinting consumables, Bioprinting nozzle
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
42142600
NACRES:
NA.25
Recommended Products
description
Microdot tips, Stainless steel, lavender, 30G, 0.25" ID 0.006" 50/box
Looking for similar products? Visit Product Comparison Guide
Application
These microdot tips are compatible with fine particle-filled materials and can produce precise micro-deposits as small as 100 μm. Burr-free, polished, passivated stainless steel dispensing tips with polypropylene SafetyLok hubs for a secure fit to barrel reservoirs.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service