Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92470

Sigma-Aldrich

2,2,4-Trimethylhexane

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)CH2C(CH3)3
CAS Number:
Molecular Weight:
128.26
Beilstein:
1718781
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.403

bp

125-127 °C (lit.)

density

0.716 g/mL at 20 °C (lit.)

SMILES string

CCC(C)CC(C)(C)C

InChI

1S/C9H20/c1-6-8(2)7-9(3,4)5/h8H,6-7H2,1-5H3

InChI key

AFTPEBDOGXRMNQ-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service