Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

924369

Sigma-Aldrich

Hydroxyapatite coating powder

≥99% trace metals basis, ≤50 ppm(Pb)

Synonym(s):

Apatite hydroxide, Ca10(PO4)6(OH)2, Hydroxyapatite, Hydroxylapatite, Pentacalcium hydroxide triphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(Ca5(OH)(PO4)3)
CAS Number:
UNSPSC Code:
12352305
NACRES:
NA.23

Quality Level

Assay

≥99% trace metals basis

size

35-45 μm

impurities

≤50 ppm (Pb)

color

white to off-white

General description

Hydroxyapatite Coating Powder consists of rounded particles obtained by spray drying. This pure and 100% synthetic HA material has been specially developed for plasma coating purposes. The specific morphology and particle size distribution enables good flowability of the powder and enables us to manufacture smooth-finished and highly adhesive coatings.

Application

Suggested use for manufacturing of inks for additive manufacturing, composite materials, and CaP based putties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sahar Vahabzadeh et al.
Acta biomaterialia, 17, 47-55 (2015-02-02)
In this work we have investigated the effects of strontium (Sr) dopant on in vitro protein release kinetics and in vivo osteogenic properties of plasma sprayed hydroxyapatite (HA) coatings, along with their dissolution behavior. Plasma sprayed HA coatings are widely
Sophie C Cox et al.
Materials science & engineering. C, Materials for biological applications, 47, 237-247 (2014-12-11)
A systematic characterisation of bone tissue scaffolds fabricated via 3D printing from hydroxyapatite (HA) and poly(vinyl)alcohol (PVOH) composite powders is presented. Flowability of HA:PVOH precursor materials was observed to affect mechanical stability, microstructure and porosity of 3D printed scaffolds. Anisotropic
Lu Xie et al.
Materials science & engineering. C, Materials for biological applications, 59, 1007-1015 (2015-12-15)
The ideal bone tissue engineering scaffolds are long-cherished with the properties of interconnected macroporous structures, adjustable degradation and excellent biocompatibility. Here, a series of porous α/β-tricalcium phosphate (α/β-TCP) biphasic bioceramics with different phase ratios of α-TCP and β-TCP were successfully

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service