Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

923761

Sigma-Aldrich

HumaTein essential bioink

derived from primary human neonatal dermal fibroblast cell culture, suitable for 3D bioprinting applications

Synonym(s):

3D bioprinting, 3DBP, ECM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NA.23

Quality Level

composition

Bioink lyophilized powder

impurities

<0.12 EU/mg Endotoxin

storage temp.

−20°C

Application

HumaTein essential bioink is based on whole ECM derived from primary human neonatal dermal fibroblast (NDFB) culture, and it’s suitable for 3D bioprinting applications. Whole ECM consists of more than 300 components includes collagen family, laminin, fibronectin, glycosaminoglycans and reserves more than 500 cytokines in balanced composition that regulate cell phenotype and function for tissue homeostasis in development. HumaTein Essential provides a rich ECM environment to promote cellular proliferation. HumaTein Essential contains human extracellular matrix components including collagens, laminin, fibronectin, tenascin, elastin, and a number of proteoglycans and glycosaminoglycans.

The cell culture derived human whole ECM effectively recapitulates tissue micro-environment during development and in organotypic cell culture. It promotes cell growth and migration and has been shown to support the proliferation of many cell types, including human embryonic stem cells, neural stem cells, endothelial cells, MSC, pericytes, fibroblasts, hepatocytes, skeletal muscle cell, smooth muscle cell, and keratinocytes. This cell culture derived human whole ECM is not denatured nor chloroform sterilized. It provides for a more tissue-like environment, due to the 3D folding structure preserved ECM proteins, which provides scaffolding structure for cell attachment, growth and migration.

Legal Information

HumaTein is a trademark of Rokit Healthcare, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin Dzobo et al.
International journal of molecular sciences, 20(18), 4628-4628 (2019-09-22)
The promise of regenerative medicine and tissue engineering is founded on the ability to regenerate diseased or damaged tissues and organs into functional tissues and organs or the creation of new tissues and organs altogether. In theory, damaged and diseased
Yuanheng Yang et al.
Acta biomaterialia, 69, 71-82 (2018-01-11)
Mesenchymal stem cell derived extracellular matrix (MSC-ECM) is a natural biomaterial with robust bioactivity and good biocompatibility, and has been studied as a scaffold for tissue engineering. In this investigation, we tested the applicability of using decellularized human bone marrow

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service