Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

917001

Sigma-Aldrich

End caps for cartridges

3 mL, blue, 50/box

Synonym(s):

3d bioprinting consumables

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NA.23

Quality Level

description

End caps (50/box), blue
Suitable for 3mL cartridge

Application

End caps feature a precision fit and use a convenient push-button to produce a snug, air-tight seal. End caps and tip caps provide an airtight seal that allows you to prefill cartridges or seal partially used syringes between shifts.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service