Skip to Content
MilliporeSigma
All Photos(1)

Documents

916358

Sigma-Aldrich

Poly(3-hydroxybutyric acid)

average Mn 10,000

Synonym(s):

P3HB, PHA, PHB, Polyhydroxybutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H6O2)nC4H6O2
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

Quality Level

mol wt

average Mn 10,000

color

white to off-white

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Poly(3-hydroxybutyric acid) (PHB) is a common polyhydroxyalkanoate (PHA), it has been widely used for food packaging, medical implants, biodegradable sutures and drug delivery systems due to its biocompatible and biodegradable properties. PHB has been investigated as drug delivery micro- and nano-carriers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Senthilkumar Pachiyappan et al.
IET nanobiotechnology, 13(4), 416-427 (2019-06-07)
In this study, polyhydroxybutyrate (PHB) nanoparticles were synthesised following nanoprecipitation method having different solvents and surfactant (Tween 80) concentrations. In this study, PHB nanoparticles were encapsulated with curcumin and subjected for sustained curcumin delivery. Both the curcumin loaded and unloaded
K Juni et al.
Critical reviews in therapeutic drug carrier systems, 3(3), 209-232 (1987-01-01)
Poly(hydroxy acids) so far have been examined for use in drug delivery in limited number, while the advantageous use of the polymers has been recognized due to their biodegradability and biocompatibility. Homo- and copolymers of lactic acid and glycolic acid
Sumeet Kapoor et al.
International journal of pharmaceutics, 511(2), 876-889 (2016-08-06)
Faster biodegradation, immunogenicity and lack of cell penetrative capabilities are hurdles in development of peptidyl drugs for cancer therapy. Polymeric carriers can be used to overcome these problems. The present study is focused on the use of polyhydroxybutyrate as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service