Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

915009

Sigma-Aldrich

Poly(L-lactide) dimethacrylate

average Mn 5,000

Synonym(s):

PLA dimethacrylate, PLA-dimethacrylate, PLLA dimethacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H4O2)nC12H18O5
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

Quality Level

mol wt

average Mn 5,000

color

off-white to beige

storage temp.

−20°C

Application

Poly(L-lactide) is a biodegradable and biocompatible polymer. It has been extensively used for tissue engineering and drug delivery research. Poly(L-lactide) dimethacrylate is functionalized with photoreactive end groups, which allows its applications in crosslinking reactions.

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Properties and medical applications of polylactic acid:A review
Express Polymer Letters, 9, 435-455 (2015)
Properties and medical applications of polylactic acid:A review
Hamad K, et al.
Express Polymer Letters, 9, 435-455 (2015)
Kai Niu et al.
Frontiers in pharmacology, 9, 930-930 (2018-08-30)
A stable doxorubicin (DOX)-loaded stereocomplex micelle drug delivery system was developed via the stereocomplex interaction between enantiomeric 4-armed poly(ethylene glycol)-poly(D-lactide) and poly(ethylene glycol)-poly(L-lactide) to realize control drug release and improve tumor cell uptake for efficient cervical carcinoma therapy. All these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service