Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

91415

Sigma-Aldrich

(Triisopropylsiloxy)methyl chloride

≥95.0% (GC)

Synonym(s):

(Chloromethoxy)triisopropylsilane, TOMCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H23ClOSi
CAS Number:
Molecular Weight:
222.83
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

form

liquid

density

0.96 g/mL at 20 °C (lit.)

functional group

chloro

SMILES string

CC(C)[Si](OCCl)(C(C)C)C(C)C

InChI

1S/C10H23ClOSi/c1-8(2)13(9(3)4,10(5)6)12-7-11/h8-10H,7H2,1-6H3

InChI key

SYZFQLUYDBIQCR-UHFFFAOYSA-N

Other Notes

Fluoride-labile 2′-O-protecting group for RNA oligonucleotide synthesis

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. Eppacher et al.
Helvetica Chimica Acta, 87, 3004-3004 (2004)
S. Pitsch et al.
Helvetica Chimica Acta, 82, 1753-1753 (1999)
S. Pitsch et al.
Helvetica Chimica Acta, 84, 3773-3773 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service