Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

913979

Sigma-Aldrich

Biotin-Thalidomide

≥95%

Synonym(s):

N1-(4-(2-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetamido)butyl)-N5-(15-oxo-19-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-4,7,10-trioxa-14-azanonadecyl)glutaramide, Biotin-conjugated thalidomide, bio-Thal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H64N8O13S
CAS Number:
Molecular Weight:
945.09
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

ligand

thalidomide

Quality Level

Assay

≥95%

form

powder

storage temp.

−20°C

SMILES string

O=C(C(CC1)N(C2=O)C(C3=C2C=CC=C3OCC(NCCCCNC(CCCC(NCCCOCCOCCOCCCNC(CCCC[C@H]4[C@](NC(N5)=O)([H])[C@]5([H])CS4)=O)=O)=O)=O)=O)NC1=O

InChI

1S/C44H64N8O13S/c53-34(12-2-1-11-33-40-30(28-66-33)49-44(61)51-40)46-19-7-21-62-23-25-64-26-24-63-22-8-20-47-36(55)14-6-13-35(54)45-17-3-4-18-48-38(57)27-65-32-10-5-9-29-39(32)43(60)52(42(29)59)31-15-16-37(56)50-41(31)58/h5,9-10,30-31,33,40H,1-4,6-8,11-28

InChI key

UCTZYEDSVDZJQR-NTZPKYJMSA-N

Application

Biotin-Thalidomide (bio-Thal) is a cereblon affinity probe for PROTAC® and targeted protein degradation research. Bio-Thal can be used in cereblon (CRBN) binding and displacement assays for protein degrader validation experiments, CRBN ligand discovery, competition analyses, or where enrichment of CRBN complexes is required.

Automate your Biotin tagging with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Behnam Nabet et al.
Nature chemical biology, 14(5), 431-441 (2018-03-28)
Dissection of complex biological systems requires target-specific control of the function or abundance of proteins. Genetic perturbations are limited by off-target effects, multicomponent complexity, and irreversibility. Most limiting is the requisite delay between modulation to experimental measurement. To enable the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service