Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

913332

Sigma-Aldrich

Boron nitride nanotubes

Multiwalled, powder, >70%

Synonym(s):

BNNT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BN
CAS Number:
UNSPSC Code:
12352300
NACRES:
NA.23

Quality Level

Assay

>70%

form

nanotubes
powder

SMILES string

B=N

InChI

1S/BN/c1-2

InChI key

PZNSFCLAULLKQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Boron nitride nanotubes (BNNT) exhibit giant stark effect and electric field tunability of the band gap. BNNTs find applications in photovoltaics, nanoelectronics and as additive in polymeric and metallic composites. BNNTs show non-cytotoxicity which makes them promising for biomedical, therapeutic and diagnostics applications as well.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boron Nitride Nanotubes for Drug Delivery.
Nanjwade B K,et al.
Advances in Materials Science Research, 10, 307-325 (2011)
Polymer Composites Reinforced by Nanotubes as Scaffolds for Tissue Engineering.
Wang W, et al.
International Journal of Polymer Science, 805634-805634 (2014)
Polymer composites of boron nitride nanotubes and nanosheets.
Meng W, et al.
Journal of Material Chemistry C, 2, 10049-10061 (2014)

Articles

Boron nitride nanotubes (BNNT) are close structural analogs of carbon nanotubes (CNT), which are high aspect ratio nanotubular material, where carbon atoms are alternately substituted by nitrogen and boron atoms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service