Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

911178

Sigma-Aldrich

PS-750-M

greener alternative

solution, 3 wt. % in H2O

Synonym(s):

FI-750-M solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H4O)nC18H33NO3
CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.22

form

liquid

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

3 wt. % in H2O

refractive index

n/D 1.3373

density

0.9931 g/mL

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced to increase catalytic efficiency and also acts as an environmentally benign and sustainable amphiphile. Click here for more information.

Application

PS-750-M 3% solution in water is a custom surfactant developed in the Handa lab.
PS-750-M allows for a variety of reactions, including challenging cross-couplings and monofluorination of indoles, to be conducted in water.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Micelle-Enabled Palladium Catalysis for Convenient sp2-sp3 Coupling of Nitroalkanes with Aryl Bromides in Water Under Mild Conditions.
Brals J, et al.
ACS Catalysis, 7245-7250 (2017)
Micelle-Enabled Palladium Catalysis for Convenient sp2-sp3 Coupling of Nitroalkanes with Aryl Bromides in Water Under Mild Conditions.
Brals J, et al.
ACS Catalysis, 7(10), 7245-7250 (2017)
Pranjal P Bora et al.
ChemSusChem, 12(13), 3037-3042 (2019-03-06)
Highly selective direct monofluorination of indoles and arenes was developed through an approach that allows site-specific solubility of substrate and fluorine source in the micelle. This approach was highly selective for a broad range of substrates with excellent functional group
Lucie Finck et al.
The Journal of organic chemistry, 83(14), 7366-7372 (2018-02-10)
Using micelles of FI-750-M, visible light, photocatalysts, and inexpensive halogenating reagents, such as N-bromosuccinimide and N-chlorosuccinimde, selective oxyhalogenations of alkynes were achieved in water under very mild conditions. No halogenation at the aromatic rings was detected, and control experiments revealed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service