909440
Fluorinated VHL spy molecule 3
≥95%
Synonym(s):
(2S,4R)-4-Hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)-1-(2,2,2-trifluoroacetyl)pyrrolidine-2-carboxamide
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
ligand
VH032 derivative
Assay
≥95%
form
powder or crystals
SMILES string
O=C([C@@H]1C[C@@H](O)CN1C(C(F)(F)F)=O)NCC2=CC=C(C3=C(C)N=CS3)C=C2
Application
Fluorinated VHL Spy Molecule 3 can be used for the discovery of new von Hippel-Lindau (VHL) binders. Based on VH032 (cat# 901490), this reporter was designed in the lab of Alessio Ciulli to bind the hydroxyproline (Hyp) recognition site of VHL. When used in competition ligand-observed 19F NMR screening experiments, its displacement indicates binding by another molecule. Additional Fluorinated VHL Spy Molecules are available as listings 909416, 909432, and 909459.
The E3 ligase VHL is of growing interest for targeted protein degradation, and these spy molecules will facilitate the identification of novel VHL ligands and VHL-based degraders.
The E3 ligase VHL is of growing interest for targeted protein degradation, and these spy molecules will facilitate the identification of novel VHL ligands and VHL-based degraders.
Other Notes
Spy vs. spy: selecting the best reporter for 19F NMR competition experiments
Structure-guided design and optimization of small molecules targeting the protein-protein interaction between the von Hippel-Lindau (VHL) E3 ubiquitin ligase and the hypoxia inducible factor (HIF) alpha subunit with in vitro nanomolar affinities
Structure-guided design and optimization of small molecules targeting the protein-protein interaction between the von Hippel-Lindau (VHL) E3 ubiquitin ligase and the hypoxia inducible factor (HIF) alpha subunit with in vitro nanomolar affinities
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of medicinal chemistry, 57(20), 8657-8663 (2014-08-29)
E3 ubiquitin ligases are attractive targets in the ubiquitin-proteasome system, however, the development of small-molecule ligands has been rewarded with limited success. The von Hippel-Lindau protein (pVHL) is the substrate recognition subunit of the VHL E3 ligase that targets HIF-1α
Chemical communications (Cambridge, England), 55(10), 1482-1485 (2019-01-16)
Systematic characterization of a series of fluorinated VHL ligands, varying binding affinity and position of the trifluoromethyl group, qualifies a spy molecule for competitive 19F NMR screening and reveals guiding principles to develop highly sensitive assays with low material consumption.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service