Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90795

Sigma-Aldrich

Tributyl borate

≥99.0% (T)

Synonym(s):

Boric acid tributylester, Tributoxyborane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3O]3B
CAS Number:
Molecular Weight:
230.15
Beilstein:
1703865
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor density

7.95 (vs air)

Assay

≥99.0% (T)

form

liquid

refractive index

n20/D 1.409 (lit.)
n20/D 1.409

bp

114-115 °C/12 mmHg (lit.)
230-235 °C (lit.)

mp

−70 °C (lit.)

density

0.853 g/mL at 25 °C (lit.)

SMILES string

CCCCOB(OCCCC)OCCCC

InChI

1S/C12H27BO3/c1-4-7-10-14-13(15-11-8-5-2)16-12-9-6-3/h4-12H2,1-3H3

InChI key

LGQXXHMEBUOXRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tributyl borate is an organoboron compound, that is often used as a reagent in organic synthesis for the preparation of various boronic acid derivatives, which are widely used in Suzuki coupling reactions in the synthesis of biaryls and other organic molecules. It is also used as a catalyst in a range of organic reactions, including the transesterification of esters and Biginelli reactions.

Application

Tributyl borate can be used as:    
  • A catalyst in the synthesis of dihydropyrimidinone derivatives via the Biginelli reaction.   
  • A boron oxide source to prepare carbon nanotube-B2O3 nanocomposite by solvothermal method.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service