Skip to Content
MilliporeSigma
All Photos(1)

Documents

907154

Sigma-Aldrich

Graphite anode powder

Synonym(s):

EL-I-C6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C
CAS Number:
UNSPSC Code:
12352302
NACRES:
NA.23

description

Advanced natural graphite
BET surface area: 3.16 m2/g or 1.52 m2/g
Calendar density: > 1.5 g/cm3
Spherical particles
Tap density: >1.0 g/cm3

form

powder

particle size

15-30 μm

cation traces

C: >99.95% (LOI)

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Application

This graphite product gives identical performance compared to commercial anode powder (@ C/2)

Electrochemistry performance:
  • Reversible capacity: 355 mAh/g
  • Irreversible capacity: 23 mAh/g
  • First cycle efficiency: 93.7%.

High Precision Coulometry (HPC)
  • Standard 2.2 Ah 18650 Cells
  • Coulombic efficiency > 0.998 at 40 Celsius

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service